Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246362.2 | XP_006246424.1 | 11 | 1424 | ||
XM_006246363.2 | XP_006246425.1 | 12 | 1530 | |||
XM_006246364.2 | XP_006246426.1 | 11 | 1451 | |||
XM_006246365.2 | XP_006246427.1 | 10 | 1331 | |||
GenBank mRNA | FQ233211.1 | - | 6 | 464 |