Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Caln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077201.1 | NP_001070669.1 | 3 | 289 | ||
XM_006249243.2 | XP_006249305.1 | 3 | 256 | |||
GenBank mRNA | DQ914829.1 | - | 2 | 163 | ||
DQ914830.1 | - | 3 | 289 |