Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC498465 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006251668.2 | XP_006251730.1 | 5 | 1299 | ||
XM_006251670.2 | XP_006251732.1 | 6 | 1174 |