Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1qtnf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191891.1 | NP_001178820.1 | 3 | 871 | ||
XM_006252134.2 | XP_006252196.1 | 4 | 1651 |