Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6V0E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003945.3 | NP_003936.1 | 3 | 303 | ||
GenBank mRNA | AK312068.1 | - | 3 | 267 | ||
BC119714.1 | - | 3 | 278 | 40117211 | ||
BC119715.1 | - | 3 | 283 | 40117212 | ||
BF691381.1 | - | 3 | 303 | |||
CR456856.1 | - | 3 | 202 | |||
CR542131.1 | - | 3 | 202 | |||
Y15286.1 | - | 3 | 264 |