Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St8sia5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213628.1 | NP_998793.1 | 2 | 168 | ||
XM_006254968.2 | XP_006255030.1 | 3 | 554 | |||
GenBank mRNA | AJ699422.1 | - | 2 | 168 |