Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sall3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108892.1 | NP_001102362.1 | 1 | 447 | ||
XM_006254985.2 | XP_006255047.1 | 2 | 562 | |||
XM_006254987.2 | XP_006255049.1 | 2 | 562 |