Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108927.1 | NP_001102397.1 | 7 | 1112 | ||
XM_006231090.1 | XP_006231152.1 | 7 | 1432 |