Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppid | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004279.1 | NP_001004279.1 | 4 | 460 | ||
GenBank mRNA | BC076386.1 | - | 4 | 460 | 7108860 | |
FQ214826.1 | - | 4 | 463 |
Gene Symbol: Ppidl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750757.3 | XP_003750805.1 | 1 | 408 |