Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhx8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012219.1 | NP_001012219.1 | 7 | 876 | ||
XM_006233528.2 | XP_006233590.1 | 8 | 1142 | |||
GenBank mRNA | BC090011.1 | - | 7 | 876 | 7324565 |