Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arrdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100770.1 | NP_001094240.1 | 7 | 868 | ||
XM_006233630.2 | XP_006233692.1 | 7 | 979 | |||
XM_006233631.2 | XP_006233693.1 | 6 | 870 | |||
GenBank mRNA | BC079372.1 | - | 7 | 860 | ||
BC158871.1 | - | 7 | 868 | 7931820 |