Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234783.1 | XP_006234845.1 | 5, 6 | 800 | ||
GenBank mRNA | FQ231962.1 | - | 5, 6 | 784 |