Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006238749.2 | XP_006238811.1 | 7 | 997 | ||
XM_008764088.1 | XP_008762310.1 | 2150 | ||||
XM_008764089.1 | XP_008762311.1 | 1970 | ||||
XM_008764090.1 | XP_008762312.1 | 7 | 1184 | |||
XM_345572.8 | XP_345573.5 | 7 | 1178 | |||
GenBank mRNA | AB235197.1 | - | 1 | 32 | ||
DQ013343.1 | - | 2 | 177 | |||
DQ013344.1 | - | 2 | 177 | |||
DQ013345.1 | - | 2 | 177 |