Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wibg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108986.1 | NP_001102456.1 | 3 | 214 | ||
XM_008765032.1 | XP_008763254.1 | 3 | 667 | |||
XM_008765033.1 | XP_008763255.1 | 3 | 628 | |||
XM_008765034.1 | XP_008763256.1 | 3 | 659 | |||
XM_008765035.1 | XP_008763257.1 | 3 | 854 | |||
XM_008765037.1 | XP_008763259.1 | 4 | 722 |