Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rdh5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765056.1 | XP_008763278.1 | 4 | 1251 | ||
GenBank mRNA | FQ218828.1 | - | 4 | 818 |