Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185085.1 | NP_001172014.1 | 19, 20 | 3117 | ||
NM_001676.5 | NP_001667.4 | 19, 20 | 3099 | |||
GenBank mRNA | AK292968.1 | - | 19, 20 | 3099 | ||
AK303498.1 | - | 12, 13 | 1851 | |||
BC031609.1 | - | 19, 20 | 2951 | 5176407 | ||
U02076.1 | - | 19, 20 | 2933 |