Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIRAS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004675.3 | NP_004666.1 | 4 | 1092 | ||
XM_017002773.1 | XP_016858262.1 | 2 | 1735 | |||
GenBank mRNA | AK021882.1 | - | 1 | 859 | ||
AK096393.1 | - | 4 | 1092 | |||
BC005362.1 | - | 2 | 602 | 3686616 | ||
CR541870.1 | - | 1 | 455 | |||
CR541892.1 | - | 1 | 455 | |||
U96750.1 | - | 2 | 603 |