Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP1A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001734.1 | NP_001001734.1 | 2, 3 | 393 | ||
NM_144699.3 | NP_653300.2 | 19, 20 | 3354 | |||
XM_011509582.1 | XP_011507884.1 | 18, 19 | 2821 | |||
GenBank mRNA | AF506797.1 | - | 19, 20 | 3346 | ||
AK098076.1 | - | 9, 10 | 1231 | |||
AL833335.1 | - | 19, 20 | 3327 | |||
BC028297.1 | - | 2, 3 | 393 | |||
BC094801.1 | - | 19, 20 | 3305 | 5268907 | ||
BX647234.1 | - | 19, 20 | 3328 |