Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF1AY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004681.3 | NP_004672.2 | 4 | 387 | ||
GenBank mRNA | AF000987.1 | - | 4 | 358 | ||
BC005248.1 | - | 4 | 269 | 3933718 | ||
BF694967.1 | - | 373 | ||||
BP331805.1 | - | 4 | 387 | |||
BT007201.1 | - | 4 | 226 |