Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109021.1 | NP_001102491.1 | 4 | 529 | ||
GenBank mRNA | BC160857.1 | - | 4 | 412 | 8365281 |