Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199119.1 | NP_954550.1 | 2 | 222 | ||
XM_006240483.1 | XP_006240545.1 | 2 | 425 | |||
XM_006240484.2 | XP_006240546.1 | 2 | 657 | |||
GenBank mRNA | AY341876.1 | - | 2 | 222 | ||
BC097262.1 | - | 2 | 209 | 7442976 |