Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004685.3 | NP_004676.3 | 7 | 1102 | ||
XM_011535306.1 | XP_011533608.1 | 8 | 1162 | |||
XM_011535307.1 | XP_011533609.1 | 8 | 1162 | |||
XM_017020845.1 | XP_016876334.1 | 7 | 1048 | |||
XM_017020846.1 | XP_016876335.1 | 4 | 367 | |||
GenBank mRNA | AF072928.1 | - | 3 | 316 | ||
AF406619.1 | - | 7 | 783 | |||
AK314587.1 | - | 7 | 967 | |||
AL832017.2 | - | 7 | 1025 | |||
BC040012.1 | - | 7 | 974 | 5495564 | ||
BU177677.1 | - | 2 | 167 |