Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1410 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000784.1 | NP_001000784.1 | 78 | |||
XM_006246372.1 | XP_006246434.1 | 1 | 78 |
Gene Symbol: Olr1408 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000785.1 | NP_001000785.1 | 1 | 78 |
Gene Symbol: Olr1411 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000783.1 | NP_001000783.1 | 1 | 72 |