Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other P2RX6 SIRNA Assays ›
Gene Symbol:
P2RX6
Gene Name:
purinergic receptor P2X 6
Gene Aliases:
P2RXL1, P2X6, P2XM
Chromosome Location:
Chr.22: 21009699 - 21028013 on Build GRCh38
UniGene ID:
Hs.113275
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s17430
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: P2RX6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001159554.1
NP_001153026.1
7
633
NM_005446.3
NP_005437.2
7
689
XM_005261819.3
XP_005261876.1
7
1413
XM_011530498.2
XP_011528800.1
7
1414
XM_011530499.2
XP_011528801.1
6
1343
XM_011530500.2
XP_011528802.1
7
1336
XM_011530501.2
XP_011528803.1
6
681
XM_011530502.2
XP_011528804.1
7
1414
XM_017029066.1
XP_016884555.1
7
1414
XM_017029067.1
XP_016884556.1
6
1341
XM_017029068.1
XP_016884557.1
7
1413
XM_017029069.1
XP_016884558.1
6
1262
XM_017029070.1
XP_016884559.1
6
615
XM_017029071.1
XP_016884560.1
6
1337
XM_017029072.1
XP_016884561.1
6
570
XM_017029073.1
XP_016884562.1
6
681
XM_017029074.1
XP_016884563.1
6
534
XM_017029075.1
XP_016884564.1
6
627
XM_017029076.1
XP_016884565.1
6
1325
GenBank mRNA
AB002058.1
-
7
704
AF065385.1
-
7
703
BC033488.1
-
7
673
5176046
BC047287.1
-
7
672
5198937
BC063553.1
-
5
526
BC064805.1
-
5
485
BC109209.2
-
7
616
40006731
BI753477.1
-
5
485
CR456535.1
-
7
695
Back To Top
More Information
Panther Classification:
Molecular Function -
ligand-gated ion channel
Gene Ontology Categories:
Function(s)
Process(es)
transport
muscle contraction
signal transduction
blood coagulation
response to ATP
purinergic nucleotide receptor signaling pathway
protein homooligomerization
protein heterooligomerization
cation transmembrane transport
purinergic nucleotide receptor activity
transmembrane signaling receptor activity
extracellular ATP-gated cation channel activity
ATP binding
channel activity
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0