Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EBAG9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278938.1 | NP_001265867.1 | 7 | 709 | ||
NM_004215.4 | NP_004206.1 | 7 | 996 | |||
NM_198120.2 | NP_936056.1 | 7 | 829 | |||
XM_017013960.1 | XP_016869449.1 | 7 | 1192 | |||
GenBank mRNA | AB007619.1 | - | 7 | 877 | ||
AF006265.1 | - | 7 | 827 | |||
AY515724.1 | - | 7 | 787 | |||
AY653072.1 | - | 7 | 729 | |||
BC005249.1 | - | 7 | 907 | 3930583 | ||
BC017729.1 | - | 7 | 780 | 3453385 | ||
BC022506.1 | - | 7 | 894 | 4815654 | ||
CR456984.1 | - | 6 | 594 |