siRNA Details
Gene Symbol: Olr1734 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001119.1 | NP_001001119.1 | 1 | 809 |

More Information
Other Species Targeted


Gene Symbol: Olr1734 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001119.1 | NP_001001119.1 | 1 | 809 |