Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ufc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003709.2 | NP_001003709.1 | 4 | 524 | ||
XM_006250251.2 | XP_006250313.1 | 5 | 479 | |||
GenBank mRNA | AB186051.1 | - | 4 | 442 | ||
AB441169.1 | - | 4 | 471 | |||
BC087648.1 | - | 4 | 524 | 7310651 | ||
FQ212748.1 | - | 4 | 542 | |||
FQ213023.1 | - | 4 | 515 | |||
FQ213507.1 | - | 4 | 525 | |||
FQ216057.1 | - | 4 | 519 | |||
FQ218926.1 | - | 4 | 545 | |||
FQ219875.1 | - | 4 | 394 | |||
FQ226625.1 | - | 4 | 537 | |||
FQ232457.1 | - | 4 | 501 |