Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009490.1 | NP_001009490.1 | 1, 2 | 210 | ||
GenBank mRNA | AY196700.1 | - | 1, 2 | 210 |