Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009507.1 | NP_001009507.1 | 1 | 212 | ||
GenBank mRNA | AY510331.1 | - | 1 | 212 |
Gene Symbol: LOC100909792 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749304.2 | XP_003749352.1 | 1 | 265 |
Gene Symbol: Vom1r56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008929.1 | NP_001008929.1 | 1 | 212 | ||
GenBank mRNA | AY510280.1 | - | 1 | 212 |