Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009509.1 | NP_001009509.1 | 1 | 352 | ||
GenBank mRNA | AY510360.1 | - | 1 | 352 |
Gene Symbol: LOC100911184 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228164.2 | XP_006228226.1 | 1 | 442 |
Gene Symbol: LOC103691019 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758872.1 | XP_008757094.1 | 1 | 352 |
Gene Symbol: Vom1r28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008966.1 | NP_001008966.1 | 1 | 352 | ||
GenBank mRNA | AY510353.1 | - | 1 | 352 |