Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008927.1 | NP_001008927.1 | 1 | 478 | ||
GenBank mRNA | AY510296.1 | - | 1 | 478 |
Gene Symbol: Vom1r101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173112.2 | NP_775135.2 | 1 | 478 | ||
GenBank mRNA | AY510287.1 | - | 1 | 478 | ||
U36786.1 | - | 1 | 396 |