Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008961.1 | NP_001008961.1 | 1 | 224 | ||
GenBank mRNA | AY510357.1 | - | 1 | 224 |
Gene Symbol: Vom1r10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008958.1 | NP_001008958.1 | 1 | 224 | ||
GenBank mRNA | AY510358.1 | - | 1 | 224 |