Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMYM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005095.2 | NP_005086.2 | 21 | 3305 | ||
XM_005271328.4 | XP_005271385.1 | 21 | 3584 | |||
XM_005271331.2 | XP_005271388.1 | 18 | 2550 | |||
XM_011542421.2 | XP_011540723.1 | 21 | 3231 | |||
XM_011542423.2 | XP_011540725.1 | 22 | 3636 | |||
XM_011542425.2 | XP_011540727.1 | 19 | 2590 | |||
XM_017002803.1 | XP_016858292.1 | 22 | 3358 | |||
XM_017002804.1 | XP_016858293.1 | 22 | 3298 | |||
XM_017002805.1 | XP_016858294.1 | 17 | 2796 | |||
GenBank mRNA | AB007885.1 | - | 17 | 2425 | ||
AK309705.1 | - | 20 | 2965 | |||
BC012093.1 | - | 19 | 2884 | |||
BC127113.1 | - | 22 | 3311 | 40125110 | ||
BC127114.1 | - | 18 | 2494 | 40125115 | ||
BX647376.1 | - | 17 | 2536 | |||
BX648067.1 | - | 12 | 1599 | |||
CR936741.1 | - | 20 | 3169 |