Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012233.1 | NP_001012233.1 | 7 | 383 | ||
GenBank mRNA | AB164465.1 | - | 7 | 383 |