Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246188.2 | XP_006246250.1 | 3 | 733 | ||
XM_006246189.2 | XP_006246251.1 | 2 | 680 | |||
XM_006246193.2 | XP_006246255.1 | 3 | 729 | |||
XM_008767726.1 | XP_008765948.1 | 2 | 4217 | |||
XM_008767727.1 | XP_008765949.1 | 2 | 4213 |