Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004739.3 | NP_004730.2 | 10 | 1303 | ||
XM_017018561.1 | XP_016874050.1 | 10 | 1155 | |||
GenBank mRNA | AB016591.1 | - | 10 | 1236 | ||
AF295807.1 | - | 10 | 913 | |||
AK301569.1 | - | 10 | 1138 | |||
BC023656.2 | - | 10 | 1099 | |||
BC053650.1 | - | 10 | 1121 | 5531965 | ||
CR749481.1 | - | 8 | 824 |