Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spdye4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034153.1 | NP_001029325.1 | 2, 3 | 469 | ||
XM_008768995.1 | XP_008767217.1 | 2, 3 | 478 | |||
XM_008768996.1 | XP_008767218.1 | 2, 3 | 414 | |||
GenBank mRNA | BC105855.1 | - | 2, 3 | 469 | 7453404 |