Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006250449.2 | XP_006250511.1 | 3, 4 | 366 | ||
XM_008769889.1 | XP_008768111.1 | 3, 4 | 366 |