Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109092.1 | NP_001102562.1 | 7, 8 | 848 | ||
XM_006251468.1 | XP_006251530.1 | 7, 8 | 843 | |||
XM_006251469.2 | XP_006251531.1 | 7, 8 | 1047 |