Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myoz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109097.1 | NP_001102567.1 | 6 | 874 | ||
XM_006251655.2 | XP_006251717.1 | 6 | 921 | |||
GenBank mRNA | FQ214748.1 | - | 6 | 874 | ||
FQ214808.1 | - | 6 | 874 | |||
FQ214853.1 | - | 6 | 857 | |||
FQ215077.1 | - | 876 | ||||
FQ215190.1 | - | 6 | 876 | |||
FQ215360.1 | - | 6 | 880 | |||
FQ215382.1 | - | 5 | 710 | |||
FQ215728.1 | - | 6 | 878 | |||
FQ215901.1 | - | 6 | 878 | |||
FQ215918.1 | - | 6 | 875 | |||
FQ216019.1 | - | 6 | 914 | |||
FQ216101.1 | - | 6 | 876 | |||
FQ216115.1 | - | 6 | 875 | |||
FQ216248.1 | - | 6 | 875 | |||
FQ216396.1 | - | 6 | 858 | |||
FQ216753.1 | - | 6 | 858 | |||
FQ216842.1 | - | 6 | 858 | |||
FQ217380.1 | - | 896 | ||||
FQ224977.1 | - | 6 | 874 |