Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102547093 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006251682.2 | XP_006251744.1 | 5 | 1037 |
Gene Symbol: LOC102546376 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770586.1 | XP_008768808.1 | 5 | 1321 |
Gene Symbol: LOC102550466 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770593.1 | XP_008768815.1 | 5 | 1226 |
Gene Symbol: LOC498465 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC079195.1 | - | 5 | 1086 |