Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUMBL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289979.1 | NP_001276908.1 | 2 | 220 | ||
NM_001289980.1 | NP_001276909.1 | 2 | 104 | |||
NM_004756.4 | NP_004747.1 | 3 | 305 | |||
GenBank mRNA | AF015041.1 | - | 3 | 339 | ||
AK289398.1 | - | 3 | 297 | |||
AK299484.1 | - | 2 | 104 | |||
BC001794.1 | - | 2 | 193 | |||
BC144583.1 | - | 2 | 115 | |||
HY111715.1 | - | 3 | 306 |
Gene Symbol: LOC100130713 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK096566.1 | - | 1 | 1616 |