Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdfy4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252759.2 | XP_006252821.1 | 10 | 1856 | ||
XM_006252760.2 | XP_006252822.1 | 10 | 3397 | |||
XM_006252762.2 | XP_006252824.1 | 10 | 1857 | |||
XM_008771159.1 | XP_008769381.1 | 11 | 1940 |