Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYTH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004227.3 | NP_004218.1 | 3 | 268 | ||
GenBank mRNA | AJ005197.1 | - | 3 | 239 | ||
AJ223957.1 | - | 3 | 189 | |||
AK313728.1 | - | 3 | 284 | |||
BC028717.1 | - | 3 | 304 | 4811560 | ||
CB988199.1 | - | 3 | 300 |