Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017508.1 | NP_001017508.1 | 5, 6 | 444 | ||
XM_006253469.2 | XP_006253531.2 | 4, 5 | 7197 | |||
XM_008771404.1 | XP_008769626.1 | 4, 5 | 7283 | |||
XM_008771405.1 | XP_008769627.1 | 3, 4 | 477 | |||
XM_008771406.1 | XP_008769628.1 | 5, 6 | 7172 | |||
GenBank mRNA | BC079268.1 | - | 5, 6 | 444 | 7133664 |