Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctla2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109115.1 | NP_001102585.1 | 4 | 409 | ||
XM_006253525.2 | XP_006253587.1 | 4 | 552 | |||
GenBank mRNA | FQ220025.1 | - | 4 | 422 | ||
FQ220118.1 | - | 4 | 422 | |||
FQ220143.1 | - | 3 | 650 | |||
FQ223122.1 | - | 2 | 189 | |||
FQ223251.1 | - | 4 | 422 | |||
FQ229997.1 | - | 4 | 353 |