Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubb2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109119.1 | NP_001102589.1 | 137 | |||
XM_008771582.1 | XP_008769804.1 | 162 |