Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197244.1 | NP_001184173.1 | 3 | 340 | ||
NM_001301061.1 | NP_001287990.1 | 4 | 462 | |||
NM_001707.3 | NP_001698.2 | 3 | 340 | |||
NR_036682.1 | 3 | 323 | ||||
GenBank mRNA | AJ223979.1 | - | 3 | 219 | ||
AK290091.1 | - | 3 | 248 | |||
AL525884.3 | - | 4 | 403 | |||
AL553260.3 | - | 3 | 607 | |||
BC000956.2 | - | 3 | 299 | 3450946 | ||
BC001967.2 | - | 3 | 206 | 3461829 | ||
BC009548.1 | - | 3 | 250 | 3897805 | ||
BI603803.1 | - | 3 | 336 | |||
BI837138.1 | - | 3 | 311 | |||
BQ109412.1 | - | 3 | 272 | |||
DA688517.1 | - | 3 | 323 | |||
X89985.1 | - | 3 | 220 |