Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109149.1 | NP_001102619.1 | 1, 2 | 231 | ||
XM_006229445.2 | XP_006229507.1 | 1, 2 | 905 | |||
XM_008759554.1 | XP_008757776.1 | 1, 2 | 905 |