Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRIP12 SIRNA Assays ›
Gene Symbol:
TRIP12
Gene Name:
thyroid hormone receptor interactor 12
Gene Aliases:
TRIP-12, ULF
Chromosome Location:
Chr.2: 229763837 - 229923234 on Build GRCh38
UniGene ID:
Hs.572642
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s17809
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRIP12
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001284214.1
NP_001271143.1
16
2470
NM_001284215.1
NP_001271144.1
14
2214
NM_001284216.1
NP_001271145.1
13
1435
NM_004238.2
NP_004229.1
15
2326
XM_005246954.4
XP_005247011.1
16
2494
XM_005246955.4
XP_005247012.1
16
2493
XM_005246956.4
XP_005247013.1
16
2473
XM_005246957.4
XP_005247014.1
16
2460
XM_005246958.4
XP_005247015.1
16
2494
XM_005246960.4
XP_005247017.1
17
2407
XM_005246961.3
XP_005247018.1
15
2344
XM_005246962.4
XP_005247019.1
15
2344
XM_006712852.3
XP_006712915.1
15
2297
XM_011512180.2
XP_011510482.1
16
2447
XM_011512181.2
XP_011510483.1
16
2445
XM_011512183.2
XP_011510485.1
15
2301
XM_017005272.1
XP_016860761.1
17
2553
XM_017005273.1
XP_016860762.1
16
2420
XM_017005274.1
XP_016860763.1
16
2368
XM_017005275.1
XP_016860764.1
16
2426
XM_017005276.1
XP_016860765.1
16
2431
XM_017005277.1
XP_016860766.1
17
2571
XM_017005278.1
XP_016860767.1
16
2386
XM_017005279.1
XP_016860768.1
16
2458
XM_017005280.1
XP_016860769.1
16
2447
XM_017005281.1
XP_016860770.1
16
2447
XM_017005282.1
XP_016860771.1
17
2406
XM_017005283.1
XP_016860772.1
16
2494
XM_017005284.1
XP_016860773.1
17
2407
XM_017005285.1
XP_016860774.1
15
2331
XM_017005286.1
XP_016860775.1
15
2366
XM_017005287.1
XP_016860776.1
15
2347
XM_017005288.1
XP_016860777.1
15
2309
XM_017005289.1
XP_016860778.1
17
2407
XM_017005290.1
XP_016860779.1
17
2407
XM_017005291.1
XP_016860780.1
16
2257
XM_017005292.1
XP_016860781.1
15
2344
XM_017005293.1
XP_016860782.1
15
2344
GenBank mRNA
AK310971.1
-
15
2344
BC113891.1
-
13
1329
40083155
BC114556.1
-
16
2364
40083165
D28476.1
-
15
2256
EU489742.1
-
14
2168
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001031659, XM_008767252, XM_008767257, XM_006245379, XM_006245377, and others.
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
DNA repair
cellular response to DNA damage stimulus
embryo development
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
negative regulation of histone H2A K63-linked ubiquitination
negative regulation of double-strand break repair
ubiquitin-protein transferase activity
protein binding
ligase activity
thyroid hormone receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0